Assay Details
| Gene Symbol: Ube2u | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033773.4 | NP_001028945.2 | 7 - 8 | 906 | 101 | |
| GenBank mRNA | AK133379.1 | - | 6 - 7 | 826 | 101 | |
| BC150880.1 | - | 7 - 8 | 898 | 9087791 | 101 | |
| BC150888.1 | - | 7 - 8 | 898 | 9087799 | 101 | |
| BC172031.1 | - | 7 - 8 | 898 | 9054317 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2u
Genomic Map