Assay Details
| Gene Symbol: Trim80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033775.3 | NP_001028947.1 | 1 - 2 | 67 | 64 | |
| GenBank mRNA | AK133415.1 | - | 1 - 2 | 68 | 64 | |
| BC147608.1 | - | 1 - 2 | 85 | 9056598 | 64 | |
| BC157912.1 | - | 1 - 2 | 85 | 9088129 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim80
Genomic Map