Assay Details
| Gene Symbol: Trim80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033775.3 | NP_001028947.1 | 3 - 4 | 772 | 92 | |
| GenBank mRNA | AK133415.1 | - | 3 - 4 | 773 | 92 | |
| BC147608.1 | - | 3 - 4 | 790 | 9056598 | 92 | |
| BC157912.1 | - | 3 - 4 | 790 | 9088129 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim80
Genomic Map