Assay Details
| Gene Symbol: Trim80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033775.3 | NP_001028947.1 | 6 - 7 | 1262 | 79 | |
| GenBank mRNA | AK133415.1 | - | 6 - 7 | 1263 | 79 | |
| BC147608.1 | - | 6 - 7 | 1280 | 9056598 | 79 | |
| BC157912.1 | - | 6 - 7 | 1280 | 9088129 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim80
Genomic Map