Assay Details
| Gene Symbol: Gm5615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033783.2 | NP_001028955.1 | 2 - 3 | 273 | 151 | |
| GenBank mRNA | AK136744.1 | - | 2 - 3 | 274 | 151 | |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0800000380.rn
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981539 | NM_001033783.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981539 | AK136744.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 62486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 127838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 152933 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318770 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57389 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981539 | NM_001033783.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981539 | AK136744.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06751414 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06751434 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06751414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26425913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07482265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08062589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07934183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07681049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07820325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08183772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07990481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26425937 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592194 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592195 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592193 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592192 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900016867.mm.1 | NM_001033783.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016867.mm.1 | NM_001033783.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016867.mm.1 | TC0900002032.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161549 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161549 | - | - |
| mouse-6 v1.1 (Illumina) | GI_85702096-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161549 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161549 | - | - |

Gm5615
Genomic Map