Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700009353.mm.1 |
NR_073205.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009353.mm.1 |
TC1700000978.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009354.mm.1 |
NR_073205.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009354.mm.1 |
TC1700000978.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009351.mm.1 |
NR_073205.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009351.mm.1 |
TC1700000978.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009352.mm.1 |
NR_073205.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009352.mm.1 |
TC1700000978.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009356.mm.1 |
TC1700000978.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009355.mm.1 |
NR_073205.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009355.mm.1 |
TC1700000978.mm.1 |
MTA TC |