Assay Details
| Gene Symbol: Gm9855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037190.1 | 1 | 1001 | 79 | ||
| GenBank mRNA | AF069519.1 | - | 1 | 962 | 79 | |
| AK089915.1 | - | 1 | 1005 | 79 | ||
| Gene Symbol: Tdg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF102875.1 | - | 6 | 677 | 79 | |
| AK169768.1 | - | 8 | 1028 | 79 | ||
| BC010315.1 | - | 8 | 994 | 3595866 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm9855
Genomic Map