Assay Details
Gene Symbol: Rangrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285442.1 | NP_001272371.1 | 1 - 2 | 217 | 85 | |
NM_001285443.1 | NP_001272372.1 | 1 - 2 | 217 | 85 | ||
NM_021329.3 | NP_067304.1 | 1 - 2 | 217 | 85 | ||
GenBank mRNA | AF243512.1 | - | 1 - 2 | 188 | 85 | |
AK010288.1 | - | 1 - 2 | 217 | 85 | ||
BB618250.1 | - | 1 - 2 | 216 | 85 | ||
BC038470.1 | - | 1 - 2 | 198 | 5344659 | 85 | |
BC087921.1 | - | 1 - 2 | 182 | 6772909 | 85 | |
BY775054.1 | - | 1 - 2 | 213 | 85 | ||
CF751505.1 | - | 1 - 2 | 70 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of membrane depolarization
transport
ER to Golgi vesicle-mediated transport
nucleocytoplasmic transport
protein transport
protein exit from endoplasmic reticulum
regulation of membrane potential
positive regulation of GTPase activity
positive regulation of establishment of protein localization to plasma membrane
regulation of sodium ion transmembrane transport
positive regulation of protein localization to cell surface
regulation of sodium ion transmembrane transporter activity
guanyl-nucleotide exchange factor activity
Ran GTPase binding
protein transporter activity
sodium channel regulator activity
ion channel binding
regulation of membrane depolarization
transport
ER to Golgi vesicle-mediated transport
nucleocytoplasmic transport
protein transport
protein exit from endoplasmic reticulum
regulation of membrane potential
positive regulation of GTPase activity
positive regulation of establishment of protein localization to plasma membrane
regulation of sodium ion transmembrane transport
positive regulation of protein localization to cell surface
regulation of sodium ion transmembrane transporter activity
guanyl-nucleotide exchange factor activity
Ran GTPase binding
protein transporter activity
sodium channel regulator activity
ion channel binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175767 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964093 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119647 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146793 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964093 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175767 | - | - |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P359078 | AK035739.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P359078 | BC087921.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P359078 | NM_021329.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P359078 | AF243512.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P359078 | BC018290.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892353 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892344 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892355 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892333 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892358 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892359 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892349 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892367 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892365 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892347 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892346 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892334 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892356 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892366 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892337 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892357 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892362 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892351 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27892360 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146793 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119647 | - | - |
Whole Mouse Genome G4122A | A_51_P359078 | AK035739.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P359078 | BC087921.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P359078 | NM_021329.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P359078 | AF243512.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P359078 | BC018290.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119647 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146793 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09651119 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09349698 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09651122 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09651125 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09651134 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09349695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27892306 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09651129 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892354 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892359 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892353 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892391 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892357 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09684967 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09349695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09548777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09429211 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09508870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892394 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09508867 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892337 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09508875 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892347 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892373 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892367 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892358 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892368 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892351 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09508868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27892344 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10558912 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387220 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558913 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500684 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558911 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448740_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10387219 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10558910 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10500683 | - | - |
Mouse Genome 430 2.0 Array | 1448740_at | - | - |
Mouse Genome 430 2.0 Array | 1448740_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100027443.mm.1 | NM_001285441.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027443.mm.1 | NM_001285442.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027443.mm.1 | NM_001285443.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027443.mm.1 | NM_021329.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027443.mm.1 | TC1100003024.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100027436.mm.1 | NM_001285441.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027436.mm.1 | NM_001285442.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027436.mm.1 | NM_001285443.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027436.mm.1 | NM_021329.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027436.mm.1 | TC1100003024.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100027449.mm.1 | NM_001285441.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027449.mm.1 | NM_001285442.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027449.mm.1 | NM_001285443.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027449.mm.1 | NM_021329.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027449.mm.1 | TC1100003024.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100027439.mm.1 | NM_001285441.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027439.mm.1 | NM_001285442.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027439.mm.1 | NM_001285443.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027439.mm.1 | NM_021329.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027439.mm.1 | TC1100003024.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100027444.mm.1 | NM_001285441.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027444.mm.1 | NM_001285442.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027444.mm.1 | NM_001285443.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027444.mm.1 | NM_021329.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027444.mm.1 | TC1100003024.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100027450.mm.1 | NM_001285442.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027450.mm.1 | NM_001285443.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027450.mm.1 | NM_021329.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027450.mm.1 | TC1100003024.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100027451.mm.1 | NM_001285442.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027451.mm.1 | NM_001285443.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027451.mm.1 | NM_021329.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100027451.mm.1 | TC1100003024.mm.1 | MTA TC |
Mouse Expression Set 430 | 1448740_PM_at | - | - |
Mouse Expression Set 430 | 1448740_at | - | - |
Murine Genome U74v2 Set | 100562_PM_at | - | - |
Murine Genome U74v2 Set | 100562_at | - | - |
Mouse Genome 430A 2.0 Array | 1448740_at | - | - |
Mouse Genome 430A 2.0 Array | 1448740_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448740_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448740_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106860400 | - | - |
MouseWG-6 v2 Expression BeadChip | 5270520 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5270520 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106860400 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005270520 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707147 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1257083 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2707147 | - | - |
mouse-6 v1.1 (Illumina) | scl057785.2_9-S | - | - |
mouse-6 v1.1 (Illumina) | GI_10946639-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5270520 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106860400 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl057785.2_9-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270520 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106860400 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2707147 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2707147 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl057785.2_9-S | - | - |
