Assay Details
| Gene Symbol: Ppih | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110129.1 | NP_001103599.1 | 1 - 2 | 109 | 81 | |
| NM_001110130.1 | NP_001103600.1 | 1 - 2 | 109 | 81 | ||
| NM_028677.4 | NP_082953.1 | 1 - 2 | 109 | 81 | ||
| GenBank mRNA | AK003179.1 | - | 1 - 2 | 87 | 81 | |
| AK005202.1 | - | 1 - 2 | 70 | 81 | ||
| AK008394.1 | - | 1 - 2 | 87 | 81 | ||
| AK014665.1 | - | 1 - 2 | 70 | 81 | ||
| AK134995.1 | - | 1 - 2 | 89 | 81 | ||
| AK136330.1 | - | 1 - 2 | 68 | 81 | ||
| AK164299.1 | - | 1 - 2 | 74 | 81 | ||
| BC016565.1 | - | 1 - 2 | 64 | 81 | ||
| BC050116.1 | - | 1 - 2 | 63 | 6432881 | 81 | |
| BC098490.1 | - | 65 | 30022355 | 81 | ||
| BY708305.1 | - | 1 - 2 | 86 | 81 | ||
| CJ071503.1 | - | 1 - 2 | 132 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppih
Genomic Map