Assay Details
Gene Symbol: Gm5431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024230.2 | NP_001019401.2 | 5 | 2133 | 83 | |
GenBank mRNA | AK144287.1 | - | 5 | 2134 | 83 | |
AK165745.1 | - | 5 | 2134 | 83 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991812 | AK165745.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991812 | NM_001024230.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991812 | AK144287.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991811 | AK165745.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991811 | NM_001024230.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991811 | AK144287.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955087 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6553 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154424 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25118 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346091 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 857828 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169747 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991811 | AK165745.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991811 | NM_001024230.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991811 | AK144287.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991812 | AK165745.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991812 | NM_001024230.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991812 | AK144287.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27780543 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27780547 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27780542 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27780549 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27780550 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27780552 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27780526 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07334403 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07334363 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07334477 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07334479 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07334385 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07334452 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07334427 | - | - |
Whole Mouse Genome G4122A | A_52_P176865 | AK165745.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P176865 | NM_001024230.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P176865 | AK144287.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07334385 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07334363 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07334427 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07334479 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09518093 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09518100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27780472 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780526 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09490290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09120933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09109990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09008181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09490289 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09056710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09093980 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09109989 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09595385 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09095361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27780514 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10385532 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10385516 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10385531 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10385530 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10385515 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10385505 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10385517 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10385504 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10385526 | - | - |
Affymetrix Mouse Clariom S Array | PSR1100024163.mm.1 | NM_001024230.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100024163.mm.1 | NM_001024230.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100024163.mm.1 | TC1100002642.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100024164.mm.1 | NM_001024230.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100024164.mm.1 | NM_001024230.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100024164.mm.1 | TC1100002642.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162671 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1248122 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3162671 | - | - |
mouse-6 v1.1 (Illumina) | GI_70608096-I | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3162671 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3162671 | - | - |
