Assay Details
Gene Symbol: Fzd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010246.1 | NP_034376.1 | 1 | 2016 | 113 | |
GenBank mRNA | AK021164.1 | - | 1 | 2200 | 113 | |
Y17709.1 | - | 1 | 1569 | 113 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
