Assay Details
| Gene Symbol: Ksr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114545.2 | NP_001108017.2 | 1 - 2 | 1014 | 88 | |
| GenBank mRNA | AK137433.1 | - | 1 - 2 | 1016 | 88 | |
| BC150883.1 | - | 1 - 2 | 1014 | 88 | ||
| BC150890.1 | - | 1 - 2 | 1014 | 88 | ||
| DQ531035.1 | - | 1 - 2 | 187 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ksr2
Genomic Map