Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10604509 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10604510 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10604508 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00013974.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013974.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013974.mm.1 |
TC0X00002187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013972.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013972.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013972.mm.1 |
TC0X00002187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013973.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013973.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013973.mm.1 |
TC0X00002187.mm.1 |
MTA TC |