Assay Details
| Gene Symbol: Defb43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039121.1 | NP_001034210.1 | 1 - 2 | 101 | 69 | |
| GenBank mRNA | BC132224.1 | - | 1 - 2 | 85 | 40130501 | 69 |
| BC132560.1 | - | 1 - 2 | 85 | 40130837 | 69 | |
| DQ012045.1 | - | 1 - 2 | 101 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119643 | NM_001039121.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119643 | DQ012045.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320928 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59684 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558531 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119643 | NM_001039121.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119643 | DQ012045.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08136361 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08136350 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08136361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29658785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11556159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11508616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11193803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11508618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11651914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11339223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11261990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11423296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11835185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11508623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11193814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11508615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11295552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11562635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11136661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29658746 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10415836 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415837 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10415835 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400007168.mm.1 | NM_001039121.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400007168.mm.1 | NM_001039121.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400007168.mm.1 | TC1400000891.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400007166.mm.1 | NM_001039121.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400007166.mm.1 | NM_001039121.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400007166.mm.1 | TC1400000891.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400007167.mm.1 | NM_001039121.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400007167.mm.1 | NM_001039121.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400007167.mm.1 | TC1400000891.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160578 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160578 | - | - |
| mouse-6 v1.1 (Illumina) | GI_84993766-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160578 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160578 | - | - |

Defb43
Genomic Map