Assay Details
Gene Symbol: Gm5868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024147.2 | NP_001019318.1 | 3 | 1017 | 150 | |
GenBank mRNA | AK084047.1 | - | 3 | 1018 | 150 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23792661 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23792662 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034870 | NM_001024147.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034870 | AK084047.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034870 | NM_001024147.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034870 | AK084047.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792624 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792643 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792680 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792667 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792670 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792689 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792694 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792691 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792693 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792661 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23792681 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05612656 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124932 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23792662 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23792661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23792670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23792693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23792689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23792556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23792668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23792694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23792691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23792609 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792623 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04209817 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04837280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04740801 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04832777 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792680 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792691 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792690 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792681 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792693 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792677 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792640 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04574057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792688 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04615431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792670 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04615432 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792668 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23792702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04598307 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04371403 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04740806 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10530502 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530501 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530500 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10530499 | - | - |
Affymetrix Mouse Clariom S Array | PSR0500022790.mm.1 | TC0500002577.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500022790.mm.1 | NM_001024147.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022790.mm.1 | NM_001024147.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022792.mm.1 | TC0500002577.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500022792.mm.1 | NM_001024147.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022792.mm.1 | NM_001024147.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022791.mm.1 | TC0500002577.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500022791.mm.1 | NM_001024147.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022791.mm.1 | NM_001024147.2 | Refseq |
Mouse Expression Set 430 | 1433578_at | - | - |
Mouse Expression Set 430 | 1433578_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 319545 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 100580484 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 100580484 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1259444 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 100580484 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100580484 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2896113 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2896113 | - | - |
