Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100017374.mm.1 |
NM_011448.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017374.mm.1 |
NM_011448.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017374.mm.1 |
TC1100001853.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017375.mm.1 |
NM_011448.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017375.mm.1 |
NM_011448.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017375.mm.1 |
TC1100001853.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017373.mm.1 |
NM_011448.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017373.mm.1 |
NM_011448.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017373.mm.1 |
TC1100001853.mm.1 |
MTA TC |