Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700017459.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017459.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017459.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017467.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017467.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017467.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017470.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017470.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017470.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017453.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017453.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017453.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017471.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017471.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017471.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017457.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017457.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017457.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017461.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017461.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017461.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017463.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017463.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017463.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017472.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017472.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017472.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017474.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017474.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017474.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017454.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017454.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017454.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017460.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017460.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017460.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003643.mm.1 |
NM_053173.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003643.mm.1 |
NM_053173.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003643.mm.1 |
TC1700000424.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017458.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017458.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017458.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003644.mm.1 |
NM_053173.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003644.mm.1 |
NM_053173.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003644.mm.1 |
TC1700000424.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017464.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017464.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017464.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017465.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017465.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017465.mm.1 |
TC1700001881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017468.mm.1 |
NM_001195298.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017468.mm.1 |
NM_001195298.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017468.mm.1 |
TC1700001881.mm.1 |
MTA TC |