Assay Details
| Gene Symbol: Tor2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152800.3 | NP_690013.1 | 2 - 3 | 475 | 73 | |
| NR_045485.1 | 2 - 3 | 389 | 73 | |||
| GenBank mRNA | AK162719.1 | - | 2 - 3 | 419 | 73 | |
| AK170979.1 | - | 1 - 2 | 1654 | 73 | ||
| AK171247.1 | - | 2 - 3 | 427 | 73 | ||
| BC003466.1 | - | 2 - 3 | 460 | 2649388 | 73 | |
| BC024469.1 | - | 2 - 3 | 438 | 4976022 | 73 | |
| CK031473.1 | - | 2 - 3 | 336 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK170979.1 transcripts.
Set Membership: |
|

Tor2a
Genomic Map