Assay Details
| Gene Symbol: Tor2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152800.3 | NP_690013.1 | 3 - 4 | 652 | 74 | |
| NR_045485.1 | 3 - 4 | 566 | 74 | |||
| GenBank mRNA | AK162719.1 | - | 3 - 4 | 596 | 74 | |
| AK170979.1 | - | 2 - 3 | 1831 | 74 | ||
| AK171247.1 | - | 3 - 4 | 604 | 74 | ||
| BC003466.1 | - | 3 - 4 | 637 | 2649388 | 74 | |
| BC023085.1 | - | 1 - 2 | 151 | 74 | ||
| BC024469.1 | - | 3 - 4 | 615 | 4976022 | 74 | |
| CK031473.1 | - | 3 - 4 | 513 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tor2a
Genomic Map