Assay Details
| Gene Symbol: Rhox3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194063.3 | NP_918952.2 | 2 - 3 | 210 | 70 | |
| GenBank mRNA | DQ058640.1 | - | 2 - 3 | 210 | 70 | |
| Gene Symbol: Rhox3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114157.1 | NP_001107629.1 | 2 - 3 | 210 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the DQ058640.1,NM_001114157.1,NM_194063.3 transcripts.
This assay is in the 5'UTR region of the DQ058640.1,NM_001114157.1,NM_194063.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984347 | NM_001114157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000417 | NM_001184964.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000417 | NM_001184969.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000417 | NR_006883.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000417 | NM_194063.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000417 | DQ058640.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P107583 | NM_194063.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984347 | NM_001114157.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P157595 | NM_001184964.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P157595 | NM_001102457.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P157595 | NM_001184969.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P157595 | NM_001040089.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P157595 | AY147207.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09265854 | - | - |
| Whole Mouse Genome G4122A | A_52_P157595 | NM_001184964.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P157595 | NM_001102457.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P157595 | NM_001184969.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P157595 | NM_001040089.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P157595 | AY147207.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107583 | NM_194063.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113419 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126216 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09265854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32323950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32323947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32323936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32323946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32323785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322389 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10604130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604134 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599252 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604133 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604132 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604133 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599252 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604132 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604134 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599253 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10604129 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10599251 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10599251 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10604129 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00002251.mm.1 | NM_194063.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002251.mm.1 | NM_194063.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002251.mm.1 | TC0X00000339.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00002249.mm.1 | NM_194063.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002249.mm.1 | NM_194063.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002249.mm.1 | TC0X00000339.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00002253.mm.1 | NM_194063.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002253.mm.1 | NM_194063.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002253.mm.1 | TC0X00000339.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013157.mm.1 | NM_001114157.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013157.mm.1 | NM_001114157.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013157.mm.1 | TC0X00003424.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013159.mm.1 | NM_001114157.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013159.mm.1 | NM_001114157.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013159.mm.1 | TC0X00003424.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00002248.mm.1 | NM_194063.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002248.mm.1 | NM_194063.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002248.mm.1 | TC0X00000339.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00002252.mm.1 | NM_194063.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002252.mm.1 | NM_194063.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002252.mm.1 | TC0X00000339.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013158.mm.1 | NM_001114157.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013158.mm.1 | NM_001114157.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013158.mm.1 | TC0X00003424.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3390333 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3390333 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003390333 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216028 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216028 | - | - |
| mouse-6 v1.1 (Illumina) | scl0378429.4_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3390333 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0378429.4_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3390333 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2928374 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2928374 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216028 | - | - |

Rhox3a
Genomic Map