Assay Details
| Gene Symbol: Tas2r129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207029.1 | NP_996912.1 | 1 | 841 | 129 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137196 | NM_207029.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117843 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176591 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137196 | NM_207029.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855399 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176591 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176591 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05624365 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05624369 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05624371 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05624376 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05624370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05624372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05517627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05630234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05408601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05489919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05823617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05408602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05624380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855400 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10542260 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10542259 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600012627.mm.1 | TC0600001569.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012627.mm.1 | NM_207029.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012627.mm.1 | NM_207029.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 301694 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 70286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000070286 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213441 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213441 | - | - |
| mouse-6 v1.1 (Illumina) | scl0387354.1_311-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 70286 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0387354.1_311-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70286 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213441 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213441 | - | - |

Tas2r129
Genomic Map