Assay Details
| Gene Symbol: Olfr427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207158.1 | NP_997041.1 | 1 | 865 | 116 | |
| GenBank mRNA | BC104103.2 | - | 1 | 865 | 40043136 | 116 |
| BC106807.2 | - | 1 | 865 | 40042912 | 116 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171136 | BC106808.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171136 | NM_207158.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171136 | BC104103.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171136 | BC104104.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171136 | BC106807.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171801 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171136 | BC106808.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171136 | NM_207158.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171136 | BC104103.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171136 | BC104104.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171136 | BC106807.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840836 | - | - |
| Whole Mouse Genome G4122A | A_52_P345128 | BC106808.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P345128 | BC104104.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103394 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00039034 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00039052 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00039045 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00039056 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00039050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00204027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00544507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00039032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00908146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00356772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00686501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00356773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00276455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00416065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840827 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351894 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351893 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 694382 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6040097 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040097 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2739797 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2739797 | - | - |
| mouse-6 v1.1 (Illumina) | scl0259162.1_92-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040097 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0259162.1_92-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040097 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2739797 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2739797 | - | - |

Olfr427
Genomic Map