Assay Details
Gene Symbol: Olfr592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207556.2 | NP_997439.2 | 1 | 439 | 119 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070466 | NM_207556.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808199 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152373 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070466 | NM_207556.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152373 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111632 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06156417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06406776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06332503 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382412 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06406775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382403 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06593906 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382419 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06035310 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06406777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06406792 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382415 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382380 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05950296 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382407 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06057548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25382432 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10555772 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10555771 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700012733.mm.1 | TC0700001483.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700012733.mm.1 | NM_207556.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700012733.mm.1 | NM_207556.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 588686 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3440736 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3440736 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003440736 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235774 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1235774 | - | - |
mouse-6 v1.1 (Illumina) | scl0404317.1_84-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3440736 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0404317.1_84-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440736 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1235774 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235774 | - | - |
