Assay Details
Gene Symbol: Gtf3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033194.2 | NP_001028366.1 | 12 - 13 | 1742 | 102 | |
GenBank mRNA | AK145040.1 | - | 1745 | 102 | ||
AK160997.1 | - | 12 - 13 | 1730 | 102 | ||
AK165830.1 | - | 12 - 13 | 1743 | 102 | ||
BC141157.1 | - | 12 - 13 | 1732 | 9055769 | 102 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
