Assay Details
| Gene Symbol: Gm694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033374.3 | NP_001028546.1 | 1 - 2 | 121 | 125 | |
| GenBank mRNA | AK145525.1 | - | 1 - 2 | 122 | 125 | |
| BC147054.1 | - | 1 - 2 | 87 | 9056056 | 125 | |
| BC147055.1 | - | 1 - 2 | 87 | 9056077 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054668 | AK145525.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562490 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054668 | AK145525.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23364052 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05430508 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03992845 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03639964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23363984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23363994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23363999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23363988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23363987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23363986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23363985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23363991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23363997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23364019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03368434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03326834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03738171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03738161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03738169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03985484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03738163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03296675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03356221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03386907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03386906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03738159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03368431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03351523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23364034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03985488 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517947 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517946 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517945 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400032129.mm.1 | TC0400003883.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400032129.mm.1 | NM_001033374.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032129.mm.1 | NM_001033374.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032131.mm.1 | TC0400003883.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400032131.mm.1 | NM_001033374.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032131.mm.1 | NM_001033374.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032128.mm.1 | TC0400003883.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400032128.mm.1 | NM_001033374.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032128.mm.1 | NM_001033374.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032130.mm.1 | TC0400003883.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400032130.mm.1 | NM_001033374.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032130.mm.1 | NM_001033374.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 599966 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160727 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160727 | - | - |
| mouse-6 v1.1 (Illumina) | GI_85701867-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160727 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160727 | - | - |

Gm694
Genomic Map