Assay Details
| Gene Symbol: Cass4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080820.2 | NP_001074289.1 | 4 - 5 | 813 | 101 | |
| NM_001276423.1 | NP_001263352.1 | 4 - 5 | 813 | 101 | ||
| GenBank mRNA | AK036419.1 | - | 4 - 5 | 787 | 101 | |
| AK080960.1 | - | 4 - 5 | 787 | 101 | ||
| AK089450.1 | - | 4 - 5 | 814 | 101 | ||
| AK170726.1 | - | 4 - 5 | 813 | 101 | ||
| BC117858.1 | - | 4 - 5 | 707 | 40095182 | 101 | |
| BC117859.1 | - | 4 - 5 | 706 | 40095183 | 101 | |
| BC129975.1 | - | 4 - 5 | 704 | 40095184 | 101 | |
More Information
Set Membership: |
|

Cass4
Genomic Map