| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300011047.mm.1 |
NM_001045535.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011047.mm.1 |
NM_001045535.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011047.mm.1 |
TC1300001775.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011048.mm.1 |
NM_001045535.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011048.mm.1 |
NM_001045535.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011048.mm.1 |
TC1300001775.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011050.mm.1 |
NM_001045535.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011050.mm.1 |
NM_001045535.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011050.mm.1 |
TC1300001775.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011045.mm.1 |
NM_001045535.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011045.mm.1 |
NM_001045535.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011045.mm.1 |
TC1300001775.mm.1 |
MTA TC |