Assay Details
| Gene Symbol: Nlrp9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139925.1 | - | 3 - 4 | 1935 | 146 | |
| AK145355.1 | - | 2 - 3 | 1624 | 146 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp9a| Gene Symbol: Nlrp9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139925.1 | - | 3 - 4 | 1935 | 146 | |
| AK145355.1 | - | 2 - 3 | 1624 | 146 | ||
Set Membership: |
|