Assay Details
| Gene Symbol: Gm9733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076679.2 | NP_001070147.1 | 2 - 3 | 502 | 110 | |
| GenBank mRNA | DQ055452.1 | - | 2 - 3 | 503 | 110 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037697 | DQ055452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037697 | NM_001076679.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623417 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625403 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399442 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795468 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45024 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431049 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758811 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 162136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 170533 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4148 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037697 | DQ055452.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037697 | NM_001076679.2 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21961223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21961248 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10497346 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497347 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497348 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10497345 | - | - |

Gm9733
Genomic Map