Assay Details
| Gene Symbol: Zfp455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048204.1 | NP_001041669.1 | 4 | 1498 | 141 | |
| GenBank mRNA | AK028336.1 | - | 3 | 1343 | 141 | |
| BC010651.1 | - | 1 | 945 | 141 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005145 | NM_001048204.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005145 | AK028336.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866002 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538644 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 296722 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176832 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132328 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103015 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005145 | NM_001048204.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005145 | AK028336.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310862 | NM_001048204.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310862 | AK028336.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29107760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10602194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10762953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10762977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10470556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10762968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10638527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10882729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10470548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29107816 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10405928 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427217_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10405927 | - | - |
| Mouse Genome 430 2.0 Array | 1427217_at | - | - |
| Mouse Genome 430 2.0 Array | 1427217_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 218311_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 218311_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300005354.mm.1 | NM_001048204.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005354.mm.1 | NM_001048204.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005354.mm.1 | TC1300000819.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300005356.mm.1 | NM_001048204.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005356.mm.1 | NM_001048204.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005356.mm.1 | TC1300000819.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300005358.mm.1 | NM_001048204.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005358.mm.1 | NM_001048204.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005358.mm.1 | TC1300000819.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300005359.mm.1 | NM_001048204.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005359.mm.1 | NM_001048204.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005359.mm.1 | TC1300000819.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300005357.mm.1 | NM_001048204.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005357.mm.1 | NM_001048204.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005357.mm.1 | TC1300000819.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300005353.mm.1 | NM_001048204.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005353.mm.1 | NM_001048204.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005353.mm.1 | TC1300000819.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427217_at | - | - |
| Mouse Expression Set 430 | 1427217_PM_at | - | - |
| Murine Genome U74v2 Set | 133111_f_at | - | - |
| Murine Genome U74v2 Set | 133111_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427217_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427217_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427217_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427217_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 402104 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2428584 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2428584 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2428584 | - | - |

Zfp455
Genomic Map