Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500008455.mm.1 |
TC1500001062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008457.mm.1 |
NM_007395.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008457.mm.1 |
NM_007395.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008457.mm.1 |
TC1500001062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008458.mm.1 |
NM_007395.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008458.mm.1 |
NM_007395.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008458.mm.1 |
TC1500001062.mm.1 |
MTA TC |