Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300004624.mm.1 |
NM_001164041.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004624.mm.1 |
NM_001164041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004624.mm.1 |
NM_001164042.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004624.mm.1 |
NM_001164042.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004624.mm.1 |
NM_008541.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004624.mm.1 |
NM_008541.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004624.mm.1 |
TC1300000694.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004625.mm.1 |
NM_001164041.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004625.mm.1 |
NM_001164041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004625.mm.1 |
NM_001164042.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004625.mm.1 |
NM_001164042.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004625.mm.1 |
NM_008541.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004625.mm.1 |
NM_008541.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004625.mm.1 |
TC1300000694.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004626.mm.1 |
NM_001164041.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004626.mm.1 |
NM_001164041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004626.mm.1 |
NM_001164042.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004626.mm.1 |
NM_001164042.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004626.mm.1 |
NM_008541.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004626.mm.1 |
NM_008541.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004626.mm.1 |
TC1300000694.mm.1 |
MTA TC |