Assay Details
| Gene Symbol: Nrtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008738.2 | NP_032764.1 | 2 - 3 | 523 | 159 | |
| GenBank mRNA | AK002508.1 | - | 2 - 3 | 663 | 159 | |
| BC057993.1 | - | 2 - 3 | 504 | 5345262 | 159 | |
| U78109.1 | - | 2 - 3 | 523 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258174 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258210 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258177 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258227 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258175 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258178 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258226 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258212 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258181 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042086 | BC057993.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042086 | U78109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042086 | NM_008738.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235602 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566252 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100938 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042086 | BC057993.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042086 | U78109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042086 | NM_008738.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P463968 | BC057993.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P463968 | U78109.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P463968 | NM_008738.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P463968 | AK002508.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31258177 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100938 | - | - |
| Whole Mouse Genome G4122A | A_52_P463968 | BC057993.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463968 | U78109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463968 | NM_008738.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463968 | AK002508.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117836 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100938 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13303484 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13539440 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13503074 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13303491 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13539434 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13215270 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13313846 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258227 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258174 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258210 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258178 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258175 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258212 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258177 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31258226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31258154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13539437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13414274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13108029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13303485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13313685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13188257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13414275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13313683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13270040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13188256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13313679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13416927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31258140 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10452150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452149 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452148 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449281_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10452147 | - | - |
| Mouse Genome 430 2.0 Array | 1449281_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449281_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18188_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18188_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700021876.mm.1 | NM_008738.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021876.mm.1 | TC1700002336.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449281_at | - | - |
| Mouse Expression Set 430 | 1449281_PM_at | - | - |
| Murine Genome U74v2 Set | 93673_PM_at | - | - |
| Murine Genome U74v2 Set | 93673_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449281_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449281_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449281_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449281_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 496871 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2450551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002450551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2655498 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2655498 | - | - |
| mouse-6 v1.1 (Illumina) | scl49758.2.1_108-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2450551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49758.2.1_108-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2450551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2655498 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2655498 | - | - |

Nrtn
Genomic Map