Assay Details
| Gene Symbol: Cer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009887.2 | NP_034017.1 | 1 - 2 | 571 | 158 | |
| GenBank mRNA | AF031896.1 | - | 1 - 2 | 585 | 158 | |
| AF035579.1 | - | 1 - 2 | 544 | 158 | ||
| BC132431.1 | - | 1 - 2 | 554 | 40130708 | 158 | |
| BC145993.1 | - | 1 - 2 | 552 | 40131296 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cer1
Genomic Map