Assay Details
Gene Symbol: Cldn34c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028426.1 | NP_082702.1 | 4 | 443 | 129 | |
GenBank mRNA | AK014019.1 | - | 4 | 411 | 129 | |
AK032219.1 | - | 3 | 438 | 129 | ||
BC027572.1 | - | 4 | 443 | 1363266 | 129 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32634536 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32634533 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32634534 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32634532 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017561 | BC027572.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017561 | NM_028426.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017561 | AK014019.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017561 | AK032219.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017561 | BC027572.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017561 | NM_028426.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017561 | AK014019.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017561 | AK032219.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965939 | AK014019.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965939 | AK032219.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P321978 | BC027572.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P321978 | NM_028426.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P321978 | AK014019.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P321978 | AK032219.1 | mouse_transcriptome |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32634533 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32634536 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32634534 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32634532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32634534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32634527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32634526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32634529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32634532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32634535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32634524 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32634523 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14685303 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14685297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14675224 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14685299 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10601600 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601592 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601596 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601589 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601597 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601599 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601591 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601590 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452640_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1439279_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10601595 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10601588 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10601598 | - | - |
Mouse Genome 430 2.0 Array | 1439279_at | - | - |
Mouse Genome 430 2.0 Array | 1439279_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1452640_at | - | - |
Mouse Genome 430 2.0 Array | 1452640_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00007595.mm.1 | NM_028426.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007595.mm.1 | TC0X00001201.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00007597.mm.1 | NM_028426.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007597.mm.1 | TC0X00001201.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00007593.mm.1 | NM_028426.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007593.mm.1 | TC0X00001201.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00007596.mm.1 | NM_028426.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007596.mm.1 | TC0X00001201.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00007592.mm.1 | NM_028426.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007592.mm.1 | TC0X00001201.mm.1 | MTA TC |
Mouse Expression Set 430 | 1452640_at | - | - |
Mouse Expression Set 430 | 1452640_PM_at | - | - |
Mouse Expression Set 430 | 1439279_PM_at | - | - |
Mouse Expression Set 430 | 1439279_at | - | - |
Murine Genome U74v2 Set | 139262_PM_f_at | - | - |
Murine Genome U74v2 Set | 139262_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1452640_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1452640_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452640_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452640_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1439279_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1439279_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 102370170 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2580919 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102370170 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102370170 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2867158 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2580919 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2867158 | - | - |
