Assay Details
Gene Symbol: Hist1h2bk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175665.2 | NP_783596.1 | 1 | 96 | 87 | |
GenBank mRNA | BC147143.1 | - | 1 | 96 | 9056184 | 87 |
BC147144.1 | - | 1 | 96 | 9056197 | 87 |
Gene Symbol: Hist1h2bc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290380.1 | NP_001277309.1 | 1 | 63 | 87 | |
NM_023422.3 | NP_075911.2 | 1 | 63 | 87 | ||
GenBank mRNA | AK005191.1 | - | 1 | 58 | 87 | |
AK005407.1 | - | 1 | 58 | 87 | ||
AK011516.1 | - | 1 | 65 | 87 | ||
BC019673.1 | - | 1 | 46 | 3993954 | 87 | |
BC082774.1 | - | 1 | 46 | 87 |
Gene Symbol: Hist1h2bj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178198.2 | NP_835505.1 | 1 | 30 | 87 |
Gene Symbol: Hist1h2bq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097979.2 | NP_001091448.2 | 1 | 58 | 87 |
Gene Symbol: Hist1h2br | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110555.1 | NP_001104025.1 | 1 | 58 | 87 | |
GenBank mRNA | AK036389.1 | - | 1 | 59 | 87 | |
BC011440.1 | - | 1 | 43 | 3989862 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
