Assay Details
Gene Symbol: Maml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK040022.1 | - | 458 | 115 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | per-hexamer repeat gene 4 | Phxr4 | Sp5 | -87.2kb | Sense | Chr. 9 - 13431361 - 13432751 on NCBI Build GRCm38 |
Downstream |
predicted gene, 39290 |
Gm39290 | 11.5kb | Sense | Chr. 9 - 13531636 - 13536932 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
