Assay Details
| Gene Symbol: A630001O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041328.1 | - | 2 | 2078 | 139 | |
| AK154268.1 | - | 1 | 977 | 139 | ||
| AK172442.1 | - | 1937 | 139 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A630001O12Rik
Genomic Map