Assay Details
| Gene Symbol: C2cd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081314.2 | NP_001074783.1 | 2 | 319 | 121 | |
| GenBank mRNA | BC039571.1 | - | 2 | 318 | 1515262 | 121 |
| BP769160.1 | - | 2 | 344 | 121 | ||
| Gene Symbol: M5C1000I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046216.1 | - | 1 | 121 | ||
| AK147236.1 | - | 1 | 121 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26600872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26600871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26600873 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26600876 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054261 | BP769161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054261 | NM_001081314.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054261 | AK014341.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054261 | BC039571.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P415395 | BP769161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P415395 | NM_001081314.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P415395 | AK014341.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P415395 | BP769161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P415395 | NM_001081314.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P415395 | AK014341.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844856 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118052 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054261 | BP769161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054261 | NM_001081314.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054261 | AK014341.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054261 | BC039571.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P415395 | BP769161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P415395 | NM_001081314.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P415395 | AK014341.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P415395 | BP769161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P415395 | NM_001081314.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P415395 | AK014341.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P415395 | BP769161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P415395 | NM_001081314.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P415395 | AK014341.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P415395 | BP769161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P415395 | NM_001081314.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P415395 | AK014341.2 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07970554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07970556 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07970570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07970559 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07970555 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26600872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26600871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26600873 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26600876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26600918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07732159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07631026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07648145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08071134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07732160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08036478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07933335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07842545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07933334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08023324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07933336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07867081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07631025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07747473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07517640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08141094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07508533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07558891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26600900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07933347 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10586615 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453326_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10594747 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10586614 | - | - |
| Mouse Genome 430 2.0 Array | 1453326_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453326_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75697_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75697_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900006828.mm.1 | NM_001081314.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006828.mm.1 | NM_001081314.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006828.mm.1 | TC0900000885.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453326_PM_at | - | - |
| Mouse Expression Set 430 | 1453326_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 446574 | - | - |
| ABI Mouse Genome Survey Microarray | 582196 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244547 | - | - |

C2cd4b
Genomic Map