Assay Details
Gene Symbol: Nacad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081652.1 | NP_001075121.1 | 4 - 5 | 4392 | 123 | |
GenBank mRNA | AB182283.1 | - | 4 - 5 | 3909 | 123 | |
BC072589.1 | - | 4 - 5 | 4392 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
