Assay Details
Gene Symbol: Nacad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081652.1 | NP_001075121.1 | 7 - 8 | 4811 | 76 | |
GenBank mRNA | AB182283.1 | - | 7 - 8 | 4328 | 76 | |
AK211170.1 | - | 1 - 2 | 50 | 76 | ||
BC072589.1 | - | 7 - 8 | 4811 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406181 | NM_001081652.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406181 | AB182283.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406181 | BC072589.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406181 | NM_001081652.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406181 | AB182283.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406181 | BC072589.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849996 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574931 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618265 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52566 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406181 | NM_001081652.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406181 | AB182283.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406181 | BC072589.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406181 | NM_001081652.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406181 | AB182283.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406181 | BC072589.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P406181 | NM_001081652.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P406181 | AB182283.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P406181 | BC072589.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P406181 | NM_001081652.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P406181 | AB182283.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P406181 | BC072589.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27569317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569329 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569343 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569348 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569341 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09536444 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09346621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09536473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569396 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569387 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569357 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569367 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09323742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569318 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569373 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09324894 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09536457 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569332 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09132772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569359 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09536416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09324888 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569397 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09536404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09324895 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09604647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569369 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09324896 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569385 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09536409 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569391 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569380 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27569324 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10384191 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384188 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384190 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384189 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384187 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384185 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384184 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384186 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455509_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1440480_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10384183 | - | - |
Mouse Genome 430 2.0 Array | 1455509_at | - | - |
Mouse Genome 430 2.0 Array | 1455509_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440480_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440480_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192950_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192950_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100021232.mm.1 | NM_001081652.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100021232.mm.1 | NM_001081652.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100021232.mm.1 | TC1100002180.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100021235.mm.1 | NM_001081652.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100021235.mm.1 | NM_001081652.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100021235.mm.1 | TC1100002180.mm.1 | MTA TC |
Mouse Expression Set 430 | 1420401_PM_a_at | - | - |
Mouse Expression Set 430 | 1420401_a_at | - | - |
Mouse Expression Set 430 | 1440480_at | - | - |
Mouse Expression Set 430 | 1440480_PM_at | - | - |
Mouse Expression Set 430 | 1455509_PM_at | - | - |
Mouse Expression Set 430 | 1455509_at | - | - |
Murine Genome U74v2 Set | 92368_at | - | - |
Murine Genome U74v2 Set | 92368_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1420401_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1420401_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455509_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440480_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455509_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440480_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1236960 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2891085 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2891085 | - | - |
