Assay Details
| Gene Symbol: Klhl34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081667.2 | NP_001075136.1 | 1 | 1852 | 106 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084177 | NM_001081667.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123388 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084177 | NM_001081667.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32768130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15170317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15170315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14754190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15102452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14612829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15190168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15221302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14721669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32768104 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10602766 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602767 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602768 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602769 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10602765 | - | - |
| Mouse Genome 430 2.0 Array | 1457077_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457077_at | - | - |
| Mouse Genome 430 2.0 Array | 1440371_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440371_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00010081.mm.1 | NM_001081667.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00010081.mm.1 | TC0X00001602.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00010080.mm.1 | NM_001081667.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00010080.mm.1 | TC0X00001602.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 323910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242984 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2965737 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2965737 | - | - |

Klhl34
Genomic Map