Assay Details
| Gene Symbol: Tomt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081679.1 | NP_001075148.1 | 2 - 3 | 338 | 88 | |
| NM_001282088.1 | NP_001269017.1 | 2 - 3 | 347 | 88 | ||
| GenBank mRNA | AK158024.1 | - | 348 | 88 | ||
| DQ854743.1 | - | 2 - 3 | 338 | 88 | ||
| EU627091.1 | - | 2 - 3 | 347 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
catecholamine metabolic process
sensory perception of sound
methylation
neurotransmitter catabolic process
catecholamine catabolic process
auditory receptor cell development
methyltransferase activity
O-methyltransferase activity
catechol O-methyltransferase activity
transferase activity
catecholamine metabolic process
sensory perception of sound
methylation
neurotransmitter catabolic process
catecholamine catabolic process
auditory receptor cell development
methyltransferase activity
O-methyltransferase activity
catechol O-methyltransferase activity
transferase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121106 | DQ854743.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121106 | EU627091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121106 | NM_001081679.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121106 | AK158024.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121106 | AK135201.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562723 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243413 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178911 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121106 | DQ854743.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121106 | EU627091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121106 | NM_001081679.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121106 | AK158024.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121106 | AK135201.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25375956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25376040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05891011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25375985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05891013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06475340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06475337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06549679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376057 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10608703 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608703 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700032468.mm.1 | NM_001081679.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032468.mm.1 | NM_001081679.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032468.mm.1 | NM_001282088.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032468.mm.1 | TC0700003847.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032472.mm.1 | NM_001081679.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032472.mm.1 | NM_001081679.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032472.mm.1 | TC0700003847.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032469.mm.1 | NM_001081679.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032469.mm.1 | NM_001081679.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032469.mm.1 | NM_001282088.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032469.mm.1 | TC0700003847.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032470.mm.1 | NM_001081679.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032470.mm.1 | NM_001081679.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032470.mm.1 | NM_001282088.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032470.mm.1 | TC0700003847.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032471.mm.1 | NM_001282088.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032471.mm.1 | TC0700003847.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2418627 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243144 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2947105 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2947105 | - | - |

Tomt
Genomic Map