Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700000946.mm.1 |
TC0700000099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000946.mm.1 |
NM_001080965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000946.mm.1 |
NM_001080965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000946.mm.1 |
NM_001080966.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000946.mm.1 |
NM_001080966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000946.mm.1 |
NM_020572.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000946.mm.1 |
NM_020572.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000947.mm.1 |
TC0700000099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000947.mm.1 |
NM_001080965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000947.mm.1 |
NM_001080965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000947.mm.1 |
NM_001080966.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000947.mm.1 |
NM_001080966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000947.mm.1 |
NM_020572.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000947.mm.1 |
NM_020572.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000945.mm.1 |
TC0700000099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000945.mm.1 |
NM_001080965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000945.mm.1 |
NM_001080965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000945.mm.1 |
NM_001080966.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000945.mm.1 |
NM_001080966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000945.mm.1 |
NM_020572.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000945.mm.1 |
NM_020572.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000950.mm.1 |
TC0700000099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000950.mm.1 |
NM_001080965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000950.mm.1 |
NM_001080965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000950.mm.1 |
NM_001080966.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000950.mm.1 |
NM_001080966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000950.mm.1 |
NM_020572.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000950.mm.1 |
NM_020572.2 |
Refseq |