Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100025291.mm.1 |
NM_009720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025291.mm.1 |
NM_009720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025291.mm.1 |
TC1100002784.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025292.mm.1 |
NM_009720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025292.mm.1 |
NM_009720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025292.mm.1 |
TC1100002784.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025294.mm.1 |
NM_009720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025294.mm.1 |
NM_009720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025294.mm.1 |
TC1100002784.mm.1 |
MTA TC |