Assay Details
Gene Symbol: Txnl4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175646.3 | NP_783577.2 | 3 - 4 | 442 | 78 | |
GenBank mRNA | AK079533.1 | - | 3 - 4 | 541 | 78 | |
AK085226.1 | - | 3 - 4 | 423 | 78 | ||
BC094379.1 | - | 3 - 4 | 458 | 30541506 | 78 | |
BC132373.1 | - | 2 - 3 | 367 | 40130650 | 78 | |
BC138333.1 | - | 2 - 3 | 367 | 8861353 | 78 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
