Assay Details
| Gene Symbol: Bend3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC060240.1 | - | 4 - 5 | 4445 | 6856073 | 144 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bend3| Gene Symbol: Bend3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC060240.1 | - | 4 - 5 | 4445 | 6856073 | 144 |
Set Membership: |
|