Assay Details
| Gene Symbol: Cyp4a31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252539.1 | NP_001239468.1 | 2 - 3 | 397 | 125 | |
| GenBank mRNA | BC013476.1 | - | 2 - 3 | 348 | 4237837 | 125 |
More Information
Set Membership: |
|

Cyp4a31
Genomic Map