Assay Details
Gene Symbol: Akr1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009658.3 | NP_033788.3 | 3 - 4 | 436 | 80 | |
GenBank mRNA | AK138000.1 | - | 3 - 4 | 389 | 80 | |
AK139250.1 | - | 3 - 4 | 391 | 80 | ||
AK145392.1 | - | 3 - 4 | 392 | 80 | ||
AK149861.1 | - | 3 - 4 | 426 | 80 | ||
AK150197.1 | - | 3 - 4 | 391 | 80 | ||
AK153949.1 | - | 3 - 4 | 420 | 80 | ||
AK164682.1 | - | 3 - 4 | 391 | 80 | ||
AK170663.1 | - | 3 - 4 | 438 | 80 | ||
AK195753.1 | - | 1 - 2 | 108 | 80 | ||
AK205407.1 | - | 1 - 2 | 108 | 80 | ||
AK206515.1 | - | 1 - 2 | 108 | 80 | ||
AK207228.1 | - | 1 - 2 | 108 | 80 | ||
AK207346.1 | - | 1 - 2 | 108 | 80 | ||
AK210404.1 | - | 1 - 2 | 108 | 80 | ||
AK211155.1 | - | 1 - 2 | 108 | 80 | ||
AK211229.1 | - | 1 - 2 | 108 | 80 | ||
AK211270.1 | - | 1 - 2 | 108 | 80 | ||
AK211569.1 | - | 1 - 2 | 108 | 80 | ||
AK212385.1 | - | 1 - 2 | 108 | 80 | ||
AK212410.1 | - | 1 - 2 | 108 | 80 | ||
AK213617.1 | - | 1 - 2 | 108 | 80 | ||
AK213838.1 | - | 1 - 2 | 108 | 80 | ||
AK215229.1 | - | 1 - 2 | 80 | |||
AK217363.1 | - | 1 - 2 | 108 | 80 | ||
AK217744.1 | - | 1 - 2 | 108 | 80 | ||
AK217924.1 | - | 1 - 2 | 108 | 80 | ||
BC004725.1 | - | 3 - 4 | 383 | 3582788 | 80 | |
BC021655.1 | - | 3 - 4 | 429 | 3708927 | 80 | |
BC085310.1 | - | 3 - 4 | 368 | 6485416 | 80 | |
BY749711.1 | - | 3 - 4 | 436 | 80 | ||
D32250.1 | - | 3 - 4 | 377 | 80 | ||
L39795.1 | - | 3 - 4 | 368 | 80 | ||
U29152.1 | - | 3 - 4 | 387 | 80 |
Gene Symbol: Gm6644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037965.1 | 1 | 391 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
