Assay Details
Gene Symbol: Ddx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025860.3 | NP_080136.2 | 12 - 13 | 1944 | 86 | |
GenBank mRNA | AK019845.1 | - | 12 - 13 | 1868 | 86 | |
AK075864.1 | - | 12 - 13 | 1881 | 86 | ||
BC017160.1 | - | 12 - 13 | 1864 | 86 | ||
BC028246.1 | - | 12 - 13 | 1915 | 2582151 | 86 | |
BC103776.1 | - | 12 - 13 | 1881 | 6492142 | 86 | |
BC141230.1 | - | 12 - 13 | 1883 | 9055842 | 86 | |
BC141231.1 | - | 12 - 13 | 1883 | 9055843 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20567562 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20567559 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20567557 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20567558 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20567561 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112997 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2002702 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351681 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75232 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65805 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219852 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215145 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623178 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635089 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156991 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119747 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125595 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2002702 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567542 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567558 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567569 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567571 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567562 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567566 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567548 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567554 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567551 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567570 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567546 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567559 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567552 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567575 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567564 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567550 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567568 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567556 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20567572 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04242627 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119747 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125595 | - | - |
Whole Mouse Genome G4122A | A_51_P235291 | BC141230.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P235291 | BC141231.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P235291 | BC017160.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P235291 | BC028246.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P235291 | BC103776.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119747 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125595 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00830549 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01055034 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01055024 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20567558 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20567557 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20567562 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20567561 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20567559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20567477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567536 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567540 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567437 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00304574 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567567 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00830548 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567564 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567570 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567554 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00744988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567517 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00830549 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00242576 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567552 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567518 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567546 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567538 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01055033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567571 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00356665 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567561 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00010371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00495341 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567572 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567575 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00356664 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567533 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567548 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567550 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567569 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00418098 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01055030 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00210949 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00418094 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20567429 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10357293 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357294 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357295 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357297 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357296 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10546806 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416070_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456000_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416071_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10357292 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10546805 | - | - |
Mouse Genome 430 2.0 Array | 1456000_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1416070_a_at | - | - |
Mouse Genome 430 2.0 Array | 1416071_at | - | - |
Mouse Genome 430 2.0 Array | 1416070_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1416071_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456000_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66942_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66942_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100023956.mm.1 | NM_025860.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100023956.mm.1 | NM_025860.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100023956.mm.1 | TC0100002974.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100023957.mm.1 | NM_025860.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100023957.mm.1 | NM_025860.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100023957.mm.1 | TC0100002974.mm.1 | MTA TC |
Mouse Expression Set 430 | 1456000_PM_at | - | - |
Mouse Expression Set 430 | 1416070_PM_a_at | - | - |
Mouse Expression Set 430 | 1416071_PM_at | - | - |
Mouse Expression Set 430 | 1416071_at | - | - |
Mouse Expression Set 430 | 1456000_at | - | - |
Mouse Expression Set 430 | 1416070_a_at | - | - |
Murine Genome U74v2 Set | 160492_PM_at | - | - |
Murine Genome U74v2 Set | 160492_at | - | - |
Murine Genome U74v2 Set | 100037_at | - | - |
Murine Genome U74v2 Set | 100037_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1456000_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416071_at | - | - |
Mouse Genome 430A 2.0 Array | 1416071_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416070_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1416070_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1456000_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416071_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416070_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416070_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416071_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 879203 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6980619 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6980619 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006980619 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246999 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1246999 | - | - |
mouse-6 v1.1 (Illumina) | scl066942.2_30-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6980619 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl066942.2_30-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980619 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1246999 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246999 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066942.2_30-S | - | - |
